Order Entry
France
Orders LinkContactUsLinkComponent
Riluzole, Reference Standards

Riluzole, Reference Standards

Supplier:   USP
Riluzole, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1604348.
  • 1604337.
  • Description:
  • Riluzole Related Compound A
  • Riluzole
  • Synonym(s):
  • 4-(Trifluoromethoxy)aniline
  • CAS No.:
  • 461-82-5
  • 1744-22-5
  • Formula:
  • C₇H₆F₃NO
  • C₈H₅F₃N₂OS
  • UN No.:
  • 2941
  • 2811

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1604348.
    Riluzole Related Compound A
    Riluzole Related Compound A
    USP reference standard
    Ampule
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 150,00 €
    1 150,00 €
    1 * 25 mg
       1 150,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1604348.
    Description
    Riluzole Related Compound A
    Synonym(s)
    4-(Trifluoromethoxy)aniline
    CAS No.
    461-82-5
    Formula
    C₇H₆F₃NO
    UN No.
    2941
  • 1604337.
    Riluzole
    Riluzole
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 479,00 €
    479,00 €
    1 * 200 mg
       479,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1604337.
    Description
    Riluzole
    Synonym(s)
    CAS No.
    1744-22-5
    Formula
    C₈H₅F₃N₂OS
    UN No.
    2811
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.