Ribavirin, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1602706.
- 1602717.
- 1602740.
- Description:
- Ribavirin
- Ribavirin Related Compound A
- Ribavirin Related Compound D
- Synonym(s):
- 1-β-D-ribofuranosyl-1H-1,2,4-triazole-3-carboxylic acid
- 1H-1,2,4-triazole-3-carboxamide
- CAS No.:
- 36791-04-5
- 39925-19-4
- 3641-08-5
- Formula:
- C₈H₁₂N₄O₅
- C₈H₁₁N₃O₆
- C₃H₄N₄O
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1602706.RibavirinSpecifications:
More
Specifications
Cat. No.1602706.DescriptionRibavirinSynonym(s)CAS No.36791-04-5FormulaC₈H₁₂N₄O₅ - 1602717.Ribavirin Related Compound ARibavirin Related Compound AUSP reference standardVial15 mgLog in for availability1 * 15 mg1 180,00 €Specifications:
More
Specifications
Cat. No.1602717.DescriptionRibavirin Related Compound ASynonym(s)1-β-D-ribofuranosyl-1H-1,2,4-triazole-3-carboxylic acidCAS No.39925-19-4FormulaC₈H₁₁N₃O₆ - 1602740.Ribavirin Related Compound DRibavirin Related Compound DUSP reference standardVial15 mgLog in for availability1 * 15 mg1 120,00 €Specifications:
More
Specifications
Cat. No.1602740.DescriptionRibavirin Related Compound DSynonym(s)1H-1,2,4-triazole-3-carboxamideCAS No.3641-08-5FormulaC₃H₄N₄O



