Anti-HLA-DQB1 Rabbit Polyclonal Antibody
Catalog # BSBTA00106-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:HLA-DQB1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for HLA-DQB1 detection. Tested with WB in Human.
Type: Primary
Antigen: HLA-DQB1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human