Order Entry
France
ContactUsLinkComponent
 
Nicotine Related Compounds, Reference Standard
 :  USP
 
Nicotine Related Compounds, Reference Standard
 :  USP
 

 

  • CAS No.:
    494-52-0
  • UN No.:
    3140
  • Synonym(s):
    (S)-3-(piperidin-2-yl)pyridine
  • Description:
    Nicotine Related Compound G

 

Frequently Bought Together