Order Entry
France
ContactUsLinkComponent
Uric acid assay kit
Uric acid assay kit
Catalog # CAYM700320-96
Supplier:  Cayman Chemical
CAS Number:  
undefined
Uric acid assay kit
Catalog # CAYM700320-96
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    Uric acid assay kit, 96 wells

Specifications

About this item

Uric Acid Assay Kit is used to measure uric acid in serum, plasma and urine.

  • Assay 40 samples in duplicate
  • Assay range: 0,5 to 10 μM
  • Plate-based fluorometric measurement (ex 530 to 540 nm, em 585 to 595 nm)

Uric acid (urate) is the end product of human purine metabolism and is mainly excreted in urine. Many factors, including genetic components and acquired factors, such as obesity and alcohol consumption, influence serum uric acid concentrations. Hyperuricemia induces or facilitates gout, kidney stones, metabolic syndrome, hypertension, and renal and cardiovascular disease, while exercise-induced acute renal failure is a significant complication of renal hypouricemia.