Order Entry
France
ContactUsLinkComponent
Organic reference standards, sumatriptans, Ph. Eur., RS
Organic reference standards, sumatriptans, Ph. Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, sumatriptans, Ph. Eur., RS
Supplier:  EDQM

Specifications

  • CAS No.:
    103628-48-4
  • Formula:
    C₁₈H₂₇N₃O₆S
  • Description:
    Sumatriptan impurity mixture

Specifications

Frequently Bought Together