Order Entry
France
ContactUsLinkComponent
Impurity and API standard, mebendazole
Impurity and API standard, mebendazole
  PROOMM0882.00
 :  
Impurity and API standard, mebendazole
  PROOMM0882.00
 :  

 

  • Pk:
    250 mg
  • Standards:
    Reference standards
  • CAS No.:
    31431-39-7
  • Description:
    Mebendazole
  • Formula:
    C₁₆H₁₃N₃O₃
  • Description:
    Mebendazole