Order Entry
France
Orders LinkContactUsLinkComponent
Impurity and API standard, doxycycline hyclate

Impurity and API standard, doxycycline hyclate

Doxycycline hyclate, Reference standards, 100 mg
Catalog # PROOMM0276.00
Impurity and API standard, doxycycline hyclate
Catalog # PROOMM0276.00
CAS Number:  
Impurity and API standard, doxycycline hyclate

Doxycycline hyclate, Reference standards, 100 mg

CAS Number:  
Price
3 580,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
Specifications

Specifications

  • Pk:
    100 mg
  • Standards:
    Reference standards
  • Description:
    Doxycycline hyclate
  • Cat. No.:
    PROOMM0276.00
  • Formula:
    C₄₆H₅₈Cl₂N₄O₁₈
  • Description:
    Doxycycline Hyclate
  • CAS No.:
    24390-14-5

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.