Order Entry
France
ContactUsLinkComponent
Organic reference standards, Ciprofloxacin-D₈ hydrochloride hydrate, Vetranal™, Supelco®
Organic reference standards, Ciprofloxacin-D₈ hydrochloride hydrate, Vetranal™, Supelco®
Catalog # 32982-10MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standards, Ciprofloxacin-D₈ hydrochloride hydrate, Vetranal™, Supelco®
Catalog # 32982-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    1216659-54-9
  • Description:
    Ciprofloxacin-d8 hydrochloride hydrate, analytical standard
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₇D₈H₁₀FN₃O₃·HCl·xH₂O
  • Description:
    Ciprofloxacin-d8 hydrochloride hydrate, analytical standard

Specifications

About this item

They are used in HPLC and gas chromatography.