Fmoc-Lys(Z)-OH 98%
: Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-carbobenzoxy-L-lysine, Fmoc-Lys(Cbz)-OH, Fmoc-Lys(Z)-OH, Nalpha-Fmoc-Nepsilon-benzyloxycarbonyl-L-lysine, Nalpha-Fmoc-Nepsilon-Cbz-L-lysine
:86060-82-4
:
:C₂₉H₃₀N₂O₆
:MFCD00065662
:502.567 g/mol
:110…120 °C
:Freezer
- Environmentally Preferable:
- Purity:97,5%
- Purity method:Assay (HPLC)
Frequently Bought Together