Order Entry
France
ContactUsLinkComponent
 
Xylylazo violet I, Sigma-Aldrich®
  248266-1G
 :  
undefined
Xylylazo violet I, Sigma-Aldrich®
  248266-1G
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance (color):
    Red to purple, brown or black
  • Appearance (form):
    Crystal to powder
  • Infrared spectrum:
    Conforms to structure
  • Wavelength (In ethanol, plus 0,08 M borate buffer):
    252 - 262 nm
  • Extinction coefficient (typically):
    >28000
  • Wavelength:
    606 - 626 nm
  • Carbon:
    50,0 - 61,0%
  • Nitrogen:
    6,7 - 7,5%

 

 

 :
 :513.50 g/mol
 :M-Clarity™ quality level = MQ200
 :14936-97-1
 :C₂₅H₂₁N₃O₆S
 :MFCD00003890