Sulphuric acid 95.0-98.0% (by acidimetry) NF, FCC, J.T.Baker®
: Avantor
:7664-93-9
:
:H₂SO₄
:MFCD00064589
:98.08 g/mol
:14,08974
:∼335 °C (1013 hPa)
:3 °C
:1.84 g/cm³ (20 °C)
:Ambient
:231-639-5
:1830
:8,II
:GHS05
:GHS07
:GHS08
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Meets N.F. Requirements:-
- Meets F.C.C. Requirements:-
- GMP Manufactured Product:-
- Food GMP Manufactured Product:-
- CAUTION: For Manufacturing, processing or repackaging:95.0 - 98.0 %
- Bulk Pharmaceutical Chemical:≤ 1 ppm
- Bulk Food Chemical:≤ 0.005 %
- NF - Assay (H₂SO₄) (by acidimetry):95.0 - 98.0 %
- NF - Arsenic (As):≤1 ppm
- NF - Chloride (Cl):≤0.005 %
- NF - Identification:Passes Test
- NF - Reducing Substances:Passes Test
- NF - Residue on Ignition:≤0.005 %
- FCC - Assay (H₂SO₄) (by acidimetry):95.0 - 98.0 %
- FCC - Arsenic (As):≤1 ppm
- FCC - Chloride (Cl):≤0.005 %
- FCC - Iron (Fe):≤0.02 %
- FCC - Identification:Passes Test
- FCC - Lead (Pb):≤5 mg/kg
- FCC - Nitrate (NO₃):≤10 mg/kg
- FCC - Reducing Substances:Passes Test
- FCC - Selenium (Se):≤0.002 %
spec_BAKR9675-02 spec_BAKR9675-03 spec_BAKR9675-07 spec_BAKR9675-08 spec_BAKR9675-25
Product specification
More
Frequently Bought Together