Order Entry
France
ContactUsLinkComponent
Sodium hydrogen sulphate monohydrate, white crystals, AR®, Macron Fine Chemicals™
Corrosive
Sodium hydrogen sulphate monohydrate, white crystals, AR®, Macron Fine Chemicals™
Catalog # 7432-20
CAS Number:  
undefined
Corrosive
Sodium hydrogen sulphate monohydrate, white crystals, AR®, Macron Fine Chemicals™
Catalog # 7432-20
CAS Number:  

Specifications

  • Pack type:
    Case, Mall STAKMOR
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
  • Solubility (10 in 50):
    Passes Test
  • Chloride (Cl):
    ≤0.001 %
  • Acidity (as H₂SO₄):
    35 - 37 %
  • Arsenic (As):
    ≤0.0001 %
  • Insoluble Matter and NH₄OH Precipitate:
    ≤0.005 %
  • Calcium and Magnesium Precipitate:
    ≤0.005 %
  • Heavy Metals (as Pb):
    ≤0.0005 %
  • Trace Impurities - Iron (Fe):
    ≤0.001 %
  • Phosphate (PO₄):
    ≤0.001 %
  • Appearance (white crystals):

Specification Test Results

About this item

 :
 :138.08 g/mol
Merck Index:13,08658
Density:2.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-665-7
UN:3260
ADR:8,III
 :GHS05
CAS Number:10034-88-5
Formula:NaHSO₄·H₂O
MDL Number:MFCD00149210