Order Entry
France
ContactUsLinkComponent
Bottles, narrow neck, rectangular, HDPE, with screw caps
Bottles, narrow neck, rectangular, HDPE, with screw caps
Catalog # SEMA5808
Supplier:  SEMADENI
Bottles, narrow neck, rectangular, HDPE, with screw caps
Catalog # SEMA5808
Supplier:  SEMADENI

Specifications

  • Capacity:
    10 l
  • W×D×H:
    190x190x365 mm
  • Neck I-Ø:
    78 mm
  • Pk:
    1

Specifications

About this item

These bottles are made of black, electrically conductive or dissipative HDPE.

  • Electrical charge is not build
  • Tight screw cap carrying strap
  • No chance of ignition

Ideal for the disposal of hazardous waste.

Certifications: UN approval under 3H2 / Y1.9 / 180.
Delivery: Delivery is individually packed with instructions.