Order Entry
France
ContactUsLinkComponent
Chemists spatulas
Chemists spatulas
Catalog # 231-1254
Supplier:  HAMMACHER KARL
undefined
Chemists spatulas
Catalog # 231-1254
Supplier:  HAMMACHER KARL

Specifications

  • Description:
    Chemists spatula, blade fine ground
  • Length:
    205 mm
  • Seq:
    04
  • Pk:
    1
  • Blade length:
    105 mm

Specifications

About this item

Chemists spatula with fine ground blade made from stainless steel.