Specifications
- Antibody type:Primary
- Antigen name:HLA class I histocompatibility antigen, alpha chain G
- Antigen symbol:HLA-G
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Gene ID:3135
- Antigen synonyms:HLA class I histocompatibility antigen|DADB-15K14.8|alpha chain G|HLA-G histocompatibility antigen|b2 microglobulin|MHC-G|HLA class I molecule|HLA G antigen|MHC class I antigen G|class I|G
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
WDR79 contains six WD (tryptophan-aspartate) repeat domains found in a number of proteins that function as adaptor molecules in signal transduction and cytoskeletal organization. The WD repeat is defined by four or more repeating units of a conserved core of approximately 40 amino acids ending with tryptophan-aspartic acid (WD). WD repeats may serve as sites of protein-protein interaction for adaptor proteins and facilitate multiprotein complex formation. The function of the WDR79 protein has not been characterized, however significant and consistent single nucleotide polymorphisms in the WDR79 gene have been found to be associated with ER negative breast cancer.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: HLA-G
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat