Anti-ATP2A1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-7539R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:ATPase, Ca++ transporting, cardiac muscle, fast twitch 1
- Antigen symbol:ATP2A1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:487
- Antigen synonyms:SERCA1|ATP2A
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the translocation of calcium from the cytosol to the sarcoplasmic reticulum lumen. Contributes to calcium sequestration involved in muscular excitation/contraction.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ATP2A1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat