Anti-WDR61 Rabbit Polyclonal Antibody (Cy7®)
BOSSBS-6886R-CY7
: Bioss
- Antibody type:Primary
- Antigen name:WD Repeat Domain 61
- Antigen symbol:WDR61
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:80349
- Antigen synonyms:WD repeat containing protein 61|WD repeat domain 61|Meiotic recombination REC14 protein homolog|SKI8|WDR61_HUMAN.|WDR 61|Recombination protein REC14|REC14
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
WDR61 is a subunit of the human PAF and SKI complexes, which function in transcriptional regulation and are involved in events downstream of RNA synthesis, such as RNA surveillance (Zhu et al., 2005 [PubMed 16024656]).[supplied by OMIM, Mar 2008].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: WDR61
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat