Order Entry
France
ContactUsLinkComponent
Anti-IMPAD1 Rabbit Polyclonal Antibody (Cy7®)
Anti-IMPAD1 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-6248R-CY7
Supplier:  Bioss
Anti-IMPAD1 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-6248R-CY7
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Inositol Monophosphatase Domain Containing 1
  • Antigen symbol:
    IMPAD1
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    54928
  • Antigen synonyms:
    IMPA3|IMP-3|gPAPP|Jaws|AI451589|B230207P20|GPAPP|AL022796|1110001C20Rik|AA408880|IMP 3
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

This gene encodes a member of the inositol monophosphatase family. The encoded protein is localized to the Golgi apparatus and catalyzes the hydrolysis of phosphoadenosine phosphate (PAP) to adenosine monophosphate (AMP). Mutations in this gene are a cause of GRAPP type chondrodysplasia with joint dislocations, and a pseudogene of this gene is located on the long arm of chromosome 1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IMPAD1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat