Order Entry
France
ContactUsLinkComponent
Anti-L-Citrulline Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anti-L-Citrulline Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-10255R-A555
Supplier:  Bioss
Anti-L-Citrulline Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-10255R-A555
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    L-Citrulline
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 555
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Multiple
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

The amino acid Citrulline is required to detoxify the liver from ammonia, which is a waste product of the body from oxidation. Citrulline promotes energy and assists with the immune system. This unusual amino acid is formed in the urea cycle by the addition of carbon dioxide and ammonia to ornithine. It is then combined with aspartic acid to form arginosuccinic acid, which later is metabolized into the amino acid arginine.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: L-Citrulline
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others