Anti-SPHK2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-2653R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Sphingosine Kinase 2
- Antigen symbol:SPHK2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:56848
- Antigen synonyms:SK-2|SK 2|SPHK2|Sphingosine kinase 2|SPK-2|SPK 2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:120-155/654
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Catalyzes the phosphorylation of sphingosine to form sphingosine 1-phosphate (SPP), a lipid mediator with both intra- and extracellular functions. Also acts on D-erythro-dihydrosphingosine, D-erythro-sphingosine and L-threo-dihydrosphingosine. Binds phosphoinositides.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SPHK2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat