Order Entry
France
ContactUsLinkComponent
Anti-SPHK2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-SPHK2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-2653R-A350
Supplier:  Bioss
Anti-SPHK2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-2653R-A350
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Sphingosine Kinase 2
  • Antigen symbol:
    SPHK2
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    56848
  • Antigen synonyms:
    SK-2|SK 2|SPHK2|Sphingosine kinase 2|SPK-2|SPK 2
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    120-155/654
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Catalyzes the phosphorylation of sphingosine to form sphingosine 1-phosphate (SPP), a lipid mediator with both intra- and extracellular functions. Also acts on D-erythro-dihydrosphingosine, D-erythro-sphingosine and L-threo-dihydrosphingosine. Binds phosphoinositides.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPHK2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat