Order Entry
France
ContactUsLinkComponent
Dissolution calibration tools
Dissolution calibration tools
Catalog # QLAADEPGAG-VK
undefined
Dissolution calibration tools
Catalog # QLAADEPGAG-VK
Specifications
  • Description:
    Depth set gauge, stainless steel, Vankel style, 25 mm
  • Seq:
    03
  • Pk:
    1
Specifications
About this item
These dissolution calibration tools are suitable for most dissolution baths.
  • Mechanical calibration parameters can easily be verified by using calibration tools
  • All calibration tools are serialised
Delivery: Supplied with NIST traceable calibration certificates and user manuals.