Anti-Oct-3 Rabbit Polyclonal Antibody [clone: Q306]
Catalog # BWRLBS3359
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Oct-3
- Clonality:Polyclonal
- Clone:Q306
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 281-330 of Human Oct-3.
OCT3 (Q306) pAb detects endogenous levels of OCT3 protein.
Type: Primary
Antigen: Oct-3
Clonality: Polyclonal
Clone: Q306
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat