Anti-PGE synthase Rabbit Polyclonal Antibody [clone: I79]
Catalog # BWRLBS2846
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PGE synthase
- Clonality:Polyclonal
- Clone:I79
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 51-100 of Human PGE synthase.
PGE synthase (I79) pAb detects endogenous levels of PGE synthase protein.
Type: Primary
Antigen: PGE synthase
Clonality: Polyclonal
Clone: I79
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat