Anti-Galpha t1 Rabbit Polyclonal Antibody [clone: M104]
Catalog # BWRLBS3311
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:G alpha t1
- Clonality:Polyclonal
- Clone:M104
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 71-120 of Human Galpha t1.
G? t1 (M104) pAb detects endogenous levels of G? t1 protein.
Type: Primary
Antigen: Galpha t1
Clonality: Polyclonal
Clone: M104
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat