Anti-GABAB R1 Rabbit Polyclonal Antibody [clone: P931]
Catalog # BWRLBS2717
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:GABAB R1
- Clonality:Polyclonal
- Clone:P931
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 930-950 of Human GABAB R1.
GABAB R1 (P931) pAb detects endogenous levels of GABAB protein.
Type: Primary
Antigen: GABAB R1
Clonality: Polyclonal
Clone: P931
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat