Anti-CRF-RII Rabbit Polyclonal Antibody [clone: H112]
BWRLBS2616
- Antibody type:Primary
- Antigen symbol:CRF-RII
- Clonality:Polyclonal
- Clone:H112
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 96-85 of Human CRF-RII.
CRF-RII (H112) pAb detects endogenous levels of CRF-RII protein.
Type: Primary
Antigen: CRF-RII
Clonality: Polyclonal
Clone: H112
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat