Anti-C/EBP-delta/epsilon Rabbit Polyclonal Antibody [clone: A217]
Catalog # BWRLBS2004
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:C/EBP- epsilon/ epsilon
- Clonality:Polyclonal
- Clone:A217
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 200-250 of Human C/EBP-delta.
C/EBP-?/? (A217) pAb detects endogenous levels of C/EBP-? and C/EBP- ?protein.
Type: Primary
Antigen: C/EBP-delta/epsilon
Clonality: Polyclonal
Clone: A217
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat