Anti-uPA Rabbit Polyclonal Antibody [clone: L239]
Catalog # BWRLBS3454
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:uPA
- Clonality:Polyclonal
- Clone:L239
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 211-260 of Human uPA.
uPA (L239) pAb detects endogenous levels of uPA protein.
Type: Primary
Antigen: uPA
Clonality: Polyclonal
Clone: L239
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human