Order Entry
France
ContactUsLinkComponent
Anti-Synuclein Rabbit Polyclonal Antibody [clone: A19]
Anti-Synuclein Rabbit Polyclonal Antibody [clone: A19]
Catalog # BWRLBS1349
Fournisseur:  Bioworld Technology
Anti-Synuclein Rabbit Polyclonal Antibody [clone: A19]
Catalog # BWRLBS1349
Fournisseur:  Bioworld Technology

Spécifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Synuclein
  • Clonality:
    Polyclonal
  • Clone:
    A19
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Spécifications

À propos de cet article

Synthetic peptide, corresponding to amino acids N-terminus of Human Synuclein-α.

Synuclein-pan (A19) pAb detects endogenous levels of Synuclein-pan protein.

Type: Primary
Antigen: Synuclein
Clonality: Polyclonal
Clone: A19
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat