Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dissolution Apparatus
Accessories for dissolution baskets, QLA
Accessories for dissolution baskets, QLA
Accessories for dissolution baskets, QLA
Catalog # QLAABSKTOOL
Supplier:
QUALITY LAB ACCESSORIES
Expand image
Accessories for dissolution baskets, QLA
Catalog # QLAABSKTOOL
Supplier:
QUALITY LAB ACCESSORIES
Specifications
Description:
Basket insertion and removal tool for clip style baskets, black acetal plastic
Seq:
02
Pk:
1
Specifications
About this item
Product Details
Product Details