Order Entry
France
ContactUsLinkComponent
Freund's complete adjuvant, MP Biomedicals
Freund's complete adjuvant, MP Biomedicals
Catalog # ICNA08642851
Supplier:  MP Biomedicals
CAS Number:  
Freund's complete adjuvant, MP Biomedicals
Catalog # ICNA08642851
Supplier:  MP Biomedicals
CAS Number:  
Specifications
  • Pk:
    50 mL
  • Description:
    Freund's complete adjuvant
Specifications
About this item
Freund's complete adjuvant is made by a modification of Freund's technique, in which 25 mg of Mycobacterium is suspended in a mixture of 7,5 ml Arlacel A and 42,5 ml paraffin oil.