Order Entry
France
ContactUsLinkComponent
Wheat germ acid phosphatase, MP Biomedicals
Wheat germ acid phosphatase, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Wheat germ acid phosphatase, MP Biomedicals
Supplier:  MP Biomedicals
Certificates

Specifications

  • CAS No.:
    9001-77-8
  • Species:
    Wheat germ
  • Storage conditions:
    −20 °C
  • Form:
    Lyophilized powder
  • Enzyme name:
    Acid Phosphatase
  • pH:
    4,0 - 7,0

Specifications

Related Information

Frequently Bought Together