Bovine native Casein (from Milk), MP Biomedicals
Casein from bovine milk is a phosphoprotein. It make up approximately 80% of the total protein in bovine milk. It is proposed to be the main protective constituent in milk.
It has a wide variety of uses, from being a major component of cheese, to use as a food additive, to a binder for safety matches. As a food source, casein supplies amino acids; carbohydrates; and two inorganic elements, calcium and phosphorus.
- CAS No.:68308-23-6
- Protein/peptide type:Native
- Source:Milk
- Species:Bovine
- Storage conditions:Ambient
- Form:Powder
- Protein/peptide name:Casein
- Colour:Dark yellow
Frequently Bought Together