Amiloride hydrochloride, Millipore®
Catalog # 129876-100
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
Synonyms: Amiloride hydrochloride hydrate, 3,5-Diamino-N-carbamimidoyl-6-chloropyrazine-2-carboxamide hydrochloride, Midamor, Colectril, Amipramizide, Guanamprazine hydrochloride, N-Amidino-3,5-diamino-6-chloropyrazinecarboxamide hydrochloride, 1-(3,5-diamino-2-chloropyrazine-6-carbonyl)guanidine hydrochloride
:
:266.09 g/mol
CAS Number:2016-88-8
:17440-83-4
Formula:C₆H₈ClN₇O·HCl
MDL Number:MFCD00058197