Order Entry
France
ContactUsLinkComponent
Anti-B2M Mouse Monoclonal Antibody (Purified) [clone: 246-E9.E7]
Anti-B2M Mouse Monoclonal Antibody (Purified) [clone: 246-E9.E7]
Supplier:  Biotium
Certificates
Anti-B2M Mouse Monoclonal Antibody (Purified) [clone: 246-E9.E7]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNUB1039-100
  • BNUB1039-500
  • Antigen symbol:
  • B2M
  • B2M
  • Antigen name:
  • Beta-2-Microglobulin
  • Beta-2-Microglobulin
  • Antigen synonyms:
  • Hdcma22p|Beta 2 microglobulin|Beta chain of MHC class I molecules|B2M|Beta-2-microglobulin form pI 5.3|Beta 2 microglobin
  • Hdcma22p|Beta 2 microglobulin|Beta chain of MHC class I molecules|B2M|Beta-2-microglobulin form pI 5.3|Beta 2 microglobin
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Clone 246-E9.E7
  • Clone 246-E9.E7
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 654514
  • 654514
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,2 mg/ml
  • 0,2 mg/ml
  • Storage buffer:
  • PBS/0,05% BSA/0,05% azide
  • PBS/0,05% BSA/0,05% azide

Specifications

Frequently Bought Together