Order Entry
France
ContactUsLinkComponent
 
Anti-PYCARD Rabbit Polyclonal Antibody (DyLight® 488)
  NOVUNBP1-78977G
Anti-PYCARD Rabbit Polyclonal Antibody (DyLight® 488)
  NOVUNBP1-78977G

 

  • Antibody type:
    Primary
  • Antigen name:
    PYD and CARD domain containing
  • Antigen symbol:
    PYCARD
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 488
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Gene ID:
    29108
  • Antigen synonyms:
    ASC, ASC, TMS, TMS1, CARD5, TMS-1, ASC, Apoptosis-associated speck-like protein containing a CARD, Caspase recruitment domain-containing protein 5, hASC
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

 

 

Type: Primary
Antigen: PYCARD
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse