Anti-SLC31A1 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Rabbit Polyclonal SLC31A1/CTR1 Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Porcine, Xenopus, Zebrafish.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:250
Type: Primary
Antigen: SLC31A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- NOVUNB100-402SS
- NOVUNB100-402
- Antigen symbol:
- Solute carrier family 31, member 1
- Solute carrier family 31, member 1
- Antigen name:
- Solute Carrier Family 31 Member A1
- Solute Carrier Family 31 Member A1
- Antigen synonyms:
- COPT1|CTR1
- COPT1|CTR1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 1317
- 1317
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together