Anti-HIF-PH2 Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNB100-137B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:HIF Prolyl Hydroxylase 2
- Antigen symbol:HIF-PH2
- Clonality:Polyclonal
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:54583
- Antigen synonyms:hypoxia-inducible factor prolyl hydroxylase 2|HPH-2|egl nine homolog 1|PNAS-118|PHD2|egl nine-like protein 1|HIF prolyl hydroxylase 2|HIFPH2|HIF-PH2|prolyl hydroxylase domain-containing protein 2|ECYT3|HIF-prolyl hydroxylase 2|C1orf12|ZMYND6|HPH2|zinc finger MYND domain-containing protein 6|SM20
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Specifications
About this item
Rabbit Polyclonal HIF Prolyl Hydroxylase 2 Antibody [Biotin]. Tested Applications: Western Blot, Flow Cytometry. Tested Reactivity: Human, Rat, Mouse (-).
Type: Primary
Antigen: HIF-PH2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human