Anti-TRPM7 Rabbit Polyclonal Antibody
Catalog # BOSSBS-9044R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Transient Receptor Potential Cation Channel Subfamily M member 7
- Antigen symbol:TRPM7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:Liquid
- Gene ID:54822
- Antigen synonyms:TRPPLIK|CHAK1|LTrpC-7|2310022G15Rik|Ltrpc7|Ltpr7|CHAK|4833414K03Rik|5033407O22Rik
- Storage buffer:Aqueous buffered solution containing 100 μg/ml BSA, 50% glycerol and 0,09% sodium azide. Store at −20 °C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4 °C
- Immunogen:800-850/1865
- Purification:Purified by Protein A
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Essential ion channel and serine/threonine-protein kinase. Divalent cation channel permeable to calcium and magnesium. Has a central role in magnesium ion homeostasis and in the regulation of anoxic neuronal cell death. Involved in TNF-induced necroptosis downstream of MLKL by mediating calcium influx. The kinase activity is essential for the channel function. May be involved in a fundamental process that adjusts plasma membrane divalent cation fluxes according to the metabolic state of the cell. Phosphorylates annexin A1 (ANXA1).
Recommended Dilutions: IHC-P: 1:100 to 500; IF(IHC-P): 1:50 to 200.
Type: Primary
Antigen: TRPM7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat