Order Entry
France
ContactUsLinkComponent
 
Anti-Mitochondrial Marker Mouse Monoclonal Antibody (CF488A) [clone: MTC719]
 :  Biotium
 
Anti-Mitochondrial Marker Mouse Monoclonal Antibody (CF488A) [clone: MTC719]
 :  Biotium

 

  • Catalog No:
  • BNC880719-100
  • BNC880719-500
  • Antigen name:
  • Mitochondrial Marker
  • Mitochondrial Marker
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone MTC719
  • Clone MTC719
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together