Order Entry
France
Orders LinkContactUsLinkComponent
Disposable cuvettes
Disposable cuvettes
Supplier:   Cole-Parmer
Disposable cuvettes
Supplier:  Cole-Parmer
Certificates
Disposable cuvettes
Disposable cuvettes
2 Options Available Below
Product Details & Documents
Plastic disposable cuvettes for liquid samples in spectrophotometry.
Product Family Options
Product Information
  • Volume
    Path length
    Window material
    Availability
    Price
  • JENW83070-32
    Disposable cuvettes
    Disposable cuvettes
    2,4 – 4,5 ml
    10 mm
    Plastic
    Log in for availability
    Log in for availability
    1 * 100 Pce
    1 * 100 Pce - 133,00 €
    133,00 €
    1 * 100 Pce
       133,00 €
  • JENW83070-30
    Disposable cuvettes
    Disposable cuvettes
    2,4 – 4,5 ml
    10 mm
    Plastic
    Log in for availability
    Log in for availability
    1 * 500 Pce
    1 * 500 Pce - 354,00 €
    354,00 €
    1 * 500 Pce
       354,00 €