Order Entry
France
ContactUsLinkComponent
Rapeseed oil (from Brassica rapa)
Rapeseed oil (from Brassica rapa)
Catalog # ICNA0296005501
Supplier:  MP Biomedicals
CAS Number:  
Rapeseed oil (from Brassica rapa)
Catalog # ICNA0296005501
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Identity Test:
    Passes

Specification Test Results

About this item

 :
CAS Number:8002-13-9
MDL Number:MFCD00217791