EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥98%, white crystalline powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:67-42-5
:
Formula:C₁₄H₂₄N₂O₁₀
MDL Number:MFCD00004291
:380.35 g/mol
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Purity:≥98%
- Melting Point:235 - 245 °C
- FTIR:Conforms to Standard
Specification Test Results
Frequently Bought Together