Tylosin tartrate, pale yellow powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:74610-55-2
:1405-54-5
Formula:C₄₆H₇₇NO₁₇·C₄H₆O₆
MDL Number:MFCD00070094
:1066.20 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Identity Test :Passes
- FTIR:Conforms to Standard
- Melting Point:137 - 147°C
- Potency:Report Result
Specification Test Results
Frequently Bought Together