Protocatechuate-3,4-dioxygenase, MP Biomedicals
Inhibitors : Ag⁺, Hg²⁺, PCMB; Optimum temperature: 60 to 65 °C; Thermal stability: below 50 °C (pH 6,0; 1 h).
One unit will oxidise one µmole of protocatechuate to 3-carboxy-cis,cis-muconate per minute at pH 7,5 at 37 °C.
- CAS No.:9029-47-4
- Enzyme name:Protocatechuate-3,4-dioxygenase
Frequently Bought Together