N-(1-Naphthyl)ethylenediamine dihydrochloride, off-white powder
:1465-25-4
:
:C₁₂H₁₆Cl₂N₂
:MFCD00012556
:259.18 g/mol
:GHS07
:Ambient
- Identity Test:Passes
- FTIR:Conforms to Standard
- Melting Point:188 - 198 °C
- λmax (water):320 - 325 nm
Frequently Bought Together