Order Entry
France
ContactUsLinkComponent
Disodium hydrogen phosphate, anhydrous, white crystalline powder ACS
Disodium hydrogen phosphate, anhydrous, white crystalline powder ACS
Supplier:  MP Biomedicals
Certificates
Disodium hydrogen phosphate, anhydrous, white crystalline powder ACS
Supplier:  MP Biomedicals

Specification Test Results

  • Assay:
    ≥99.0%
  • pH (5% aq soln):
    8,7 - 9,3
  • Insoluble Matter:
    ≤0,01%
  • Loss on Drying (105 °C):
    ≤0,2%
  • Chloride (Cl):
    ≤0,002%
  • Sulphate (SO4):
    ≤0,005%
  • Heavy Metals:
    ≤0,001%
  • pH:
    ≤0,002%
  • pka:
    Iron (Fe)

Specification Test Results

Frequently Bought Together