Order Entry
France
ContactUsLinkComponent
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) ≥98%, white powder
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) ≥98%, white powder
Catalog # ICNA04808196
Supplier:  MP Biomedicals
CAS Number:  
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) ≥98%, white powder
Catalog # ICNA04808196
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Identity Test:
    Passes
  • Purity:
    ≥98%
  • FTIR:
    Conforms to Standard
  • OD 400 (5% aq.):
    <0.03
  • OD 290 (5% aq.):
    <0.10

Specification Test Results

About this item

 :
 :614.89 g/mol
Storage Temperature:Ambient
CAS Number:75621-03-3
Formula:C₃₂H₅₈N₂O₇S
MDL Number:MFCD00081083